Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFT46 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113392
Description
IFT46 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Forms part of a complex involved in intraflagellar transport (IFT), the bi-directional movement of particles required for the assembly, maintenance and functioning of primary cilia. May play a role in chondrocyte maturation and skeletogenesis.
Specifications
| IFT46 | |
| Polyclonal | |
| Unconjugated | |
| IFT46 | |
| 1500035H01Rik; AA408110; AI663846; c11orf2; C11orf60; CFAP32; cilia and flagella associated protein 32; Ift46; intraflagellar transport 46; intraflagellar transport 46 homolog; intraflagellar transport protein 46 homolog; intraflagellar transport protein IFT46; R75047; UPF0360 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 56912, 76568 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9DB07, Q9NQC8 | |
| IFT46 | |
| Recombinant Human Intraflagellar transport protein 46 homolog protein (1-304AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction