Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT52 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100068 Supplier Invitrogen™ Supplier No. PA5100068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFT52.

Forms part of a complex involved in intraflagellar transport (IFT), the bi-directional movement of particles required for the assembly, maintenance and functioning of primary cilia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT52
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFT52
Gene Accession No. Q62559, Q9Y366
Gene Alias BC037708; C20orf9; CGI-53; Ift52; intraflagellar transport 52; intraflagellar transport 52 homolog; intraflagellar transport protein 52 homolog; NGD2; NGD5; Protein NGD5; Protein NGD5 homolog; RGD1311004
Gene Symbols IFT52
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFT52(Accession Q9Y366), corresponding to amino acid residues P319-Q369.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245866, 362265, 51098
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.