Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT52 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566920 Supplier Invitrogen™ Supplier No. PA566920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPLQLIQPQFE TPLPTLQPAV FPPSFRELPP PPLELFDLDE TFSSEKARLA QITNKCTEED LEFYVRKCGD ILGVTSKLPK DQQDA Highest antigen sequence identity to the following orthologs - mouse 95%, rat 27%.

Forms part of a complex involved in intraflagellar transport (IFT), the bi-directional movement of particles required for the assembly, maintenance and functioning of primary cilia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT52
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFT52
Gene Accession No. Q9Y366
Gene Alias BC037708; C20orf9; CGI-53; Ift52; intraflagellar transport 52; intraflagellar transport 52 homolog; intraflagellar transport protein 52 homolog; NGD2; NGD5; Protein NGD5; Protein NGD5 homolog; RGD1311004
Gene Symbols IFT52
Host Species Rabbit
Immunogen Recombinant Human IFT52. Recombinant protein control fragment (Product #RP-107573).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51098
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.