Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFT57 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
SDP

Catalog No. 502542688
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2688 30 μL
50-167-2058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2688 Supplier OriGene Supplier No. TA500917S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFT57 Monoclonal antibody specifically detects IFT57 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen IFT57
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2G3
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PLEK
Gene Accession No. P08567
Gene Alias 4833420A15Rik; dermal papilla-derived protein 8; DERP8; Esrrbl1; estrogen-related receptor beta like 1; estrogen-related receptor beta-like protein 1; FLJ10147; HIP1 protein interactor; HIP1-interacting protein; Hippi; huntingtin interacting protein-1 interacting protein; huntingtin-interacting protein-1 protein interactor; IFT57; intraflagellar transport 57; intraflagellar transport 57 homolog; intraflagellar transport protein 57 homolog; MHS4R2; Vestrogen-related receptor beta like 1
Host Species Mouse
Immunogen Full length human recombinant protein of human PLEK produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5341
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.