Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT80 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528970 Supplier Invitrogen™ Supplier No. PA528970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (81%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The IFT80 gene encodes a protein with 7 WD40 domains that is a component of the intraflagellar transport. The IFT is essential for the development and maintenance of motile and sensory cilia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT80
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene IFT80
Gene Accession No. Q9P2H3
Gene Alias 4921524P20Rik; ATD2; IFT80; intraflagellar transport 80; intraflagellar transport 80 homolog; intraflagellar transport protein 80 homolog; intraflagellar transport protein 80i-like protein; KIAA1374; mKIAA1374; RGD1312006; SRTD2; WD repeat domain 56; WD repeat-containing protein 56; WDR56
Gene Symbols IFT80
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IFT80. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57560
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.