Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT80 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557439
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557439 Supplier Invitrogen™ Supplier No. PA557439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EELYSCSDDH QIVKWNLLTS ETTQIVKLPD DIYPIDFHWF PKSLGVKKQT QAESFVLTSS DGKFHLISKL GRVEKSVEAH CGAVLAGRWN Y Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The IFT80 gene encodes a protein with 7 WD40 domains that is a component of the intraflagellar transport. The IFT is essential for the development and maintenance of motile and sensory cilia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT80
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFT80
Gene Accession No. Q9P2H3
Gene Alias 4921524P20Rik; ATD2; IFT80; intraflagellar transport 80; intraflagellar transport 80 homolog; intraflagellar transport protein 80 homolog; intraflagellar transport protein 80i-like protein; KIAA1374; mKIAA1374; RGD1312006; SRTD2; WD repeat domain 56; WD repeat-containing protein 56; WDR56
Gene Symbols IFT80
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IFT80. Recombinant protein control fragment (Product #RP-96273).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57560
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.