Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFT81 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297511DL594
Description
IFT81 Polyclonal specifically detects IFT81 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IFT81 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
28981 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
carnitine deficiency-associated gene expressed in ventricle 1, Carnitine deficiency-associated protein expressed in ventricle 1, CDV-1, CDV1carnitine deficiency-associated, expressed in ventricle 1, CDV1R, CDV-1R, intraflagellar transport 81 homolog (Chlamydomonas), intraflagellar transport protein 81 homolog, MGC102777, MGC4027 | |
Produced in rabbits immunized with E. coli-derived Human IFT81 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction