Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT81 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553921 Supplier Invitrogen™ Supplier No. PA553921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVVSEPAMGH SDLLELESKI NEINTEINQL IEKKMMRNEP IEGKLSLYRQ QASIISRKKE AKAEELQEAK EKLASLEREA SVKRNQTREF DGTEV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

Isoform CDV-1 may be involved in cardiac hypertrophy caused by carnitine deficiency. Isoform CDV-1R appears to play a role in development of the testis and spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT81
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFT81
Gene Accession No. Q8WYA0
Gene Alias AW060663; carnitine deficiency-associated gene expressed in ventricle 1; carnitine deficiency-associated protein expressed in ventricle 1; Cdv1; CDV-1; CDV1R; CDV-1R; DV1; Ift81; intraflagellar transport 81; intraflagellar transport 81 homolog; intraflagellar transport protein 81 homolog
Gene Symbols IFT81
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IFT81. Recombinant protein control fragment (Product #RP-92291).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28981
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.