Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFT81 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305102100UL
Description
IFT81 Polyclonal specifically detects IFT81 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
IFT81 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
28981 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
carnitine deficiency-associated gene expressed in ventricle 1, Carnitine deficiency-associated protein expressed in ventricle 1, CDV-1, CDV1carnitine deficiency-associated, expressed in ventricle 1, CDV1R, CDV-1R, intraflagellar transport 81 homolog (Chlamydomonas), intraflagellar transport protein 81 homolog, MGC102777, MGC4027 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 550-676 of human IFT81 (NP_054774.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction