Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFT88 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588948 Supplier Thermo Scientific Supplier No. PA588948
Add to Cart
Add to Cart

IFT88 Polyclonal Antibody

IFT88 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT88
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene IFT88
Gene Accession No. Q13099, Q61371
Gene Alias AW552028; D13S1056E; DAF19; fb37b11; flexo; fxo; HGNC:20606; hTg737; Ift88; intraflagellar transport 88; intraflagellar transport 88 homolog; intraflagellar transport protein; intraflagellar transport protein 88; intraflagellar transport protein 88 homolog; MGC26259; orpk; osm-5; oval; ovl; polaris; polaris homolog; polaris; axonemal transport protein; probe hTg737 (polycystic kidney disease, autosomal recessive); recessive polycystic kidney disease protein Tg737; Recessive polycystic kidney disease protein Tg737 homolog; RP11-172H24.2; testicular tissue protein Li 93; tetratricopeptide repeat domain 10; tetratricopeptide repeat protein 10; TG737; Tg737 protein; Tg737Rpw; TgN(Imorpk)737Rpw; TgN737Rpw; TPR repeat protein 10; transgene insert site 737, insertional mutation, polycystic kidney disease; Ttc10; wu:fb37b11; xx:zah8127
Gene Symbols IFT88
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 534-833 of human IFT88 (NP_7831952).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21821, 8100
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.