Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig gamma-4 Chain C Region Antibody (IGHG4/1345), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254461AF750
Description
Ig gamma-4 Chain C Region Monoclonal specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Ig gamma-4 Chain C Region | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
IGHG4 | |
Recombinant human Ig gamma-4 Chain C Region fragment (Uniprot: P01861) | |
0.1 mL | |
3503.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
IGHG4/1345 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker) | |
Mouse | |
Protein A or G purified | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction