Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig gamma-4 Chain C Region Antibody (IGHG4/2042R), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP279850AF647
Description
Ig gamma-4 Chain C Region Monoclonal specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Ig gamma-4 Chain C Region | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHG4 | |
Recombinant full-length human Ig gamma-4 Chain C Region protein (Uniprot: P01861) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
IGHG4/2042R | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker) | |
Rabbit | |
Protein A purified | |
RUO | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction