Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig gamma-4 Chain C Region, Rabbit anti-Human, Clone: IGHG4/2042R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50 - $560.50
Specifications
Antigen | Ig gamma-4 Chain C Region |
---|---|
Clone | IGHG4/2042R |
Concentration | 0.2 mg/mL |
Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
Ig gamma-4 Chain C Region Monoclonal specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Ig gamma-4 Chain C Region | |
0.2 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Rabbit | |
Human | |
P01861 | |
3503 | |
Recombinant full-length human Ig gamma-4 Chain C Region protein | |
Primary | |
Store at 4°C. |
IGHG4/2042R | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker) | |
IGHG4 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title