Learn More
Ig Kpa Lgt Chn Mouse, anti-Rat, R718, Clone: MRK-1, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752105
Description
The MRK-1 monoclonal antibody reacts specifically with rat Igs bearing κ light chains. It does not react with rat immunoglobulin λ light chains or heavy chains. The MRK-1 antibody is effective for detection of cell-surface Ig by immunofluorescent staining with flow cytometric analysis. It has not been shown to stimulate B-cell proliferation.

Specifications
Ig Kpa Lgt Chn | |
Monoclonal | |
R718 | |
50 μg | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
MRK-1 | |
Mouse | |
RUO | |
Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.