Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig lambda light chain Mouse anti-Human, Clone: Lamb14, Abnova™

Catalog No. 89034373 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-373 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-373 Supplier Abnova Corporation Supplier No. MAB14291

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human Ig lambda light chain.

  • High specificity
  • High reactivity

Specifications

Antigen Ig lambda light chain
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone Lamb14
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human Ig lambda light chain.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determ
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P0CG04
Gene Alias IGLC
Gene Symbols IGLC1
Host Species Mouse
Immunogen Native purified human lambda light chain.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3537
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.