Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig, κ Light Chain Mouse anti-Human, BUV737, Clone: G20-193, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 749125
Description
Ig, κ Light Chain Monoclonal antibody specifically detects Ig, κ Light Chain in Human samples. It is validated for Flow Cytometry
Specifications
| Ig, κ Light Chain | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| Ig, kappa Light Chain; IGKC; HCAK1; Km; Ig kappa chain C region | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
| Flow Cytometry | |
| G20-193 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction