Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig, κ Light Chain Mouse anti-Mouse, BUV805, Clone: MRK-1, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 748627
Description
Ig, κ Light Chain Monoclonal antibody specifically detects Ig, κ Light Chain in Mouse samples. It is validated for Flow Cytometry
Specifications
| Ig, κ Light Chain | |
| Monoclonal | |
| Brilliant Ultraviolet 805 | |
| Igkc; Ig κ light chain; Ig kappa light chain | |
| Pooled rat Ig | |
| RUO | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| MRK-1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction