Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig, λ Light Chain Mouse anti-Mouse, BV480, Clone: B46-5, BD OptiBuild™

Catalog No. BDB746315 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB746315 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB746315 Supplier BD Biosciences Supplier No. 746315
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ig, λ Light Chain Monoclonal antibody specifically detects Ig, λ Light Chain in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig, λ Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone B46-5
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 480
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.