Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig, κ Light Chain Mouse anti-Rat, BB700, Clone: MRK-1, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 746031
Description
Ig, κ Light Chain Monoclonal antibody specifically detects Ig, κ Light Chain in Rat samples. It is validated for Flow Cytometry.
Specifications
Ig, κ Light Chain | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Rat | |
Purified |
Flow Cytometry | |
MRK-1 | |
Brilliant Blue 700 (BB700) | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction