Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig, κ light chain Rat anti-Mouse, BV480, Clone: 187.1, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 746752
Description
Ig, κ light chain Monoclonal antibody specifically detects Ig, κ light chain in Mouse samples. It is validated for Flow Cytometry.
Specifications
Ig, κ light chain | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
187.1 | |
Brilliant Violet 480 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction