Learn More
Ig (Rabbit) polyclonal antibody, Abnova™
Rabbit Ig can be used as Isotype Control with Rabbit MaxPab
Supplier: Abnova Corporation PAB14685E01
Specifications
| Ig | |
| Polyclonal | |
| No additive | |
| None | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| 100 μL | |
| Antibody |