Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA Antibody (7D5F12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$572.50
Specifications
Antigen | IgA |
---|---|
Clone | 7D5F12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Paraffin 1:200-1:1000 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61794
![]() |
Novus Biologicals
NBP261794 |
0.1 mL |
Each of 1 for $572.50
|
|
|||||
Description
IgA Monoclonal specifically detects IgA in Human, Mouse, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IgA | |
1 mg/mL | |
Monoclonal | |
Purified | |
Adaptive Immunity, Immunology | |
Immunoglobulin A | |
IghA1 | |
IgG1 | |
Protein G purified |
7D5F12 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Unconjugated | |
Mouse | |
Human, Mouse, Monkey | |
3493 | |
Purified recombinant fragment of human IghA1 (AA: 207-353) expressed in E. Coli. | |
Primary | |
37.6 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title