Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA Antibody (HISA43), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311510APC
Description
IgA Monoclonal antibody specifically detects IgA in Human samples. It is validated for Flow CytometrySpecifications
IgA | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
HISA43 | |
Flow Cytometry | |
Immunoglobulin A | |
Purified human IgA | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3493 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction