Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA Antibody (SPM187), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2450520.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
IgA Monoclonal antibody specifically detects IgA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
IgA | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 1 - 2 ug/million cells in 0.1ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Immunoglobulin A | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This MAb is specific to heavy chain of IgA and shows minimal cross-reaction with heavy chains of other immunoglobulins. It is reactive with both IgA1 and IgA2 subclasses of Alpha heavy chain. It reacts with the third constant domain (CH3) of the alpha chain of IgA molecules. Immunoglobulins are four-chain, Y-shaped, monomeric structures comprised of two identical heavy chains and two identical light chains held together through inter-chain disulfide bonds. The chains form two domains, the Fab (antigen binding) fragment and the Fc (constant) fragment. Immunoglobulin A (IgA) is the main protein of the mucosal immune system. It is generated by B-cells in gut-associated lymphoid tissues. Daily production of IgA exceeds that of any of the other immunoglobulins. IgA exists mainly in dimers but can also exist as polymers or as monomers. Dimers and polymers contain a joining (J) chain that can be bound by the polymeric immunoglobulin receptor (pIgR) for transportation of the molecule to mucosal surfaces. The most common feature of plasmacytomas, and certain non-Hodgkin s lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. | |
Store at 4C. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
SPM187 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
IGHA1 | |
Purified human IgA | |
0.2 mg | |
Adaptive Immunity, Immunology | |
3493 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction