Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA (α-Heavy Chain) Ab-2 Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB1433R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB1433R7 7 mL Prediluted
RB1433A1 500 μL Concentrated
RB1433A0 100 μL Concentrated
RB1433A 1 mL Concentrated
RB1433R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RB1433R7 Supplier Epredia Supplier No. RB1433R7

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

IgA (α-Heavy Chain) Ab-2, Rb PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: IgA isolated from a pool of normal human sera

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic Ab-2 recognizes a protein of 50kDa, identified as alpha heavy chain of human immunoglobulins. It reacts with both IgA1 and IgA2 isotypes. Ab-2 does not cross-react with γ (IgG), μ (IgM), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Ab-2 is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen IgA (alpha-Heavy Chain) Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen IgA isolated from a pool of normal human sera
Quantity 7 mL
Regulatory Status IVD
Research Discipline Immunoglobulins
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.