Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA Heavy Chain Antibody (IGHA/7567R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320648100UG
Description
IgA Heavy Chain Monoclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
IgA Heavy Chain | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
Rabbit | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
IGHA/7567R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Full-length native protein (purified) corresponding to Human IgA | |
100 μg | |
Primary | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction