Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA Lambda Human, HRP, Isotype Control, Novus Biologicals™
SDP

Catalog No. NB179955 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179955 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179955 Supplier Novus Biologicals Supplier No. NBP306874H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Polyclonal Isotype Control

IgA Lambda Polyclonal antibody specifically detects IgA Lambda in Human samples. It is validated for ELISA, Control

Specifications

Antigen IgA λ
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Host Species Human
Immunogen Human myeloma serum
Quantity 0.1 mL
Regulatory Status RUO
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Isotype Controls
Isotype IgA λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.