Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA Monoclonal Antibody (5-C3), Huabio

Catalog No. 502544453
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4453 Supplier HUABIO RESEARCH INC Supplier No. M151030
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgA Monoclonal antibody specifically detects IgA in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen IgA
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5-C3
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene IGHA2
Gene Accession No. P01876, P01877
Gene Alias A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker)
Host Species Mouse
Immunogen Native protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3493, 3494
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.