Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA Mouse anti-Human, DyLight 550, Clone: AD3, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500469R
Description
IgA Monoclonal specifically detects IgA in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IgA | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHA1 | |
Purified human IgA. | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3493 | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
AD3 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Immunoglobulin A | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
The antibody AD3 reacts with alpha-chain of human IgA1 and IgA2. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction