Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA1 Rat anti-Human, Clone: MT50, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP3182881000UG
This item is not returnable.
View return policy
Description
IgA1 Monoclonal antibody specifically detects IgA1 in Human samples. It is validated for ELISA, ELISPOTSpecifications
IgA1 | |
Monoclonal | |
0.5 mg/ml | |
0.2 um filtered PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
MT50 | |
Unconjugated | |
Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
Human IgA | |
1000 μg | |
Immunology, Stem Cell Markers | |
3493 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction