Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA2 Mouse anti-Human, Clone: 14AS (14A), Novus Biologicals™
SDP

Catalog No. NB002823 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002823 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002823 Supplier Novus Biologicals Supplier No. NBP268309500UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IgA2 Fc Monoclonal specifically detects IgA2 Fc in Human samples. It is validated for ELISA.

Specifications

Antigen IgA2
Applications ELISA
Classification Monoclonal
Clone 14AS (14A)
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation PBS
Gene Alias immunoglobulin heavy constant alpha 2 (A2m marker)
Gene Symbols IGHA2
Host Species Mouse
Immunogen Purified human IgA from colostrum.
Quantity 500 μg
Research Discipline Immunology
Gene ID (Entrez) 3494
Test Specificity Mouse anti Human IgA2 antibody, clone 14AS, reacts with human IgA2, Fc-region. The antibody shows about 5% cross reactivity with IgA1 and 3% with IgM.
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.