Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGBP1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100487JF646
Description
IGBP1 Polyclonal antibody specifically detects IGBP1 in Human, Mouse samples. It is validated for Western BlotSpecifications
IGBP1 | |
Polyclonal | |
Western Blot | |
ALPHA-4, B cell signal transduction molecule alpha 4, bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ), B-cell signal transduction molecule alpha 4, CD79a-binding protein 1, IBP1, immunoglobulin (CD79A) binding protein 1, immunoglobulin-binding protein 1, Protein alpha-4, protein phosphatase 2A, regulatory subunit alpha-4, Renal carcinoma antigen NY-REN-16 | |
The immunogen recognized by this antibody maps to a region between residue 300 and the C-terminus (residue 339) of human Protein Phosphatase 2A, regulatory subunit alpha-4 (Immunoglobulin binding protein 1) using the numbering given in SwissProt entry P78318 (GeneID 3476). | |
0.1 mL | |
Protein Phosphatase, Signal Transduction | |
3476 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction