Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGBP1 Antibody (OTI4F6), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271009AF594
Description
IGBP1 Monoclonal specifically detects IGBP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IGBP1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
IGBP1 | |
Full length recombinant protein of human IGBP1 (NP_001542) produced in HEK293T cells. | |
Primary | |
Human | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI4F6 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
ALPHA-4, B cell signal transduction molecule alpha 4, bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ), B-cell signal transduction molecule alpha 4, CD79a-binding protein 1, IBP1, immunoglobulin (CD79A) binding protein 1, immunoglobulin-binding protein 1, Protein alpha-4, protein phosphatase 2A, regulatory subunit alpha-4, Renal carcinoma antigen NY-REN-16 | |
Mouse | |
0.1 mL | |
3476 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction