Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGBP1 Antibody (OTI4F6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271009
Description
IGBP1 Monoclonal specifically detects IGBP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
IGBP1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready | |
ALPHA-4, B cell signal transduction molecule alpha 4, bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ), B-cell signal transduction molecule alpha 4, CD79a-binding protein 1, IBP1, immunoglobulin (CD79A) binding protein 1, immunoglobulin-binding protein 1, Protein alpha-4, protein phosphatase 2A, regulatory subunit alpha-4, Renal carcinoma antigen NY-REN-16 | |
Mouse | |
39 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
OTI4F6 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
IGBP1 | |
Full length human recombinant protein of human IGBP1 (NP_001542) produced in HEK293T cell. | |
Affinity Purified | |
Protein Phosphatase, Signal Transduction | |
3476 | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction