Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGBP1 Monoclonal Antibody (OTI4F6)
GREENER_CHOICE

Catalog No. PIMA525107
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525107 Supplier Invitrogen™ Supplier No. MA525107
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGBP1 Monoclonal Antibody for Western Blot, ICC/IF, Flow

The proliferation and differentiation of B cells is dependent upon a B-cell antigen receptor (BCR) complex. Binding of antigens to specific B-cell receptors results in a tyrosine phosphorylation reaction through the BCR complex and leads to multiple signal transduction pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGBP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4F6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IGBP1
Gene Accession No. P78318
Gene Alias AI115586; AI662457; alpha 4; ALPHA-4; Alpha4 phosphoprotein; AW208785; B cell signal transduction molecule alpha 4; bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ); B-cell signal transduction molecule alpha 4; C81413; CD79a-binding protein 1; IBP1; Igbp1; immunoglobulin (CD79A) binding protein 1; immunoglobulin binding protein 1; immunoglobulin-binding protein 1; Lymphocyte signal transduction molecule alpha 4; p52; Pc52; phosphoprotein, component, 52 kDa; protein alpha-4; Protein phosphatase 2/4/6 regulatory subunit; protein phosphatase 2A, regulatory subunit alpha-4; renal carcinoma antigen NY-REN-16; Tap42
Gene Symbols IGBP1
Host Species Mouse
Immunogen Full length human recombinant protein of IGBP1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3476
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.