Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGBP1 Monoclonal Antibody (OTI4F6), TrueMAB™, OriGene
SDP

Catalog No. 502542468
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2468 30 μL
50-167-2336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2468 Supplier OriGene Supplier No. TA501193S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGBP1 Monoclonal antibody specifically detects IGBP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen IGBP1
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone OTI4F6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Pde4a
Gene Accession No. P27815
Gene Alias AI115586; AI662457; alpha 4; ALPHA-4; Alpha4 phosphoprotein; AW208785; B cell signal transduction molecule alpha 4; bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ); B-cell signal transduction molecule alpha 4; C81413; CD79a-binding protein 1; IBP1; Igbp1; immunoglobulin (CD79A) binding protein 1; immunoglobulin binding protein 1; immunoglobulin-binding protein 1; Lymphocyte signal transduction molecule alpha 4; p52; Pc52; phosphoprotein, component, 52 kDa; protein alpha-4; Protein phosphatase 2/4/6 regulatory subunit; protein phosphatase 2A, regulatory subunit alpha-4; renal carcinoma antigen NY-REN-16; Tap42
Host Species Mouse
Immunogen Full length human recombinant protein of human PDE4A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5141
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.