Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGBP1 Mouse anti-Human, Mouse, Clone: 2D11A8, Proteintech
SDP

Catalog No. 501736353 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6353 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6353 Supplier Proteintech Group Inc Supplier No. 602561IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The proliferation and differentiation of B cells is dependent upon a B-cell antigen receptor (BCR) complex. Binding of antigens to specific B-cell receptors results in a tyrosine phosphorylation reaction through the BCR complex and leads to multiple signal transduction pathways.

Specifications

Antigen IGBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2D11A8
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IGBP1
Gene Accession No. P78318, Q61249
Gene Alias ALPHA 4, CD79a binding protein 1, IBP1, IGBP1, Protein alpha 4
Gene Symbols IGBP1
Host Species Mouse
Immunogen IGBP1 Fusion Protein Ag6782
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18518, 3476
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.