Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529897
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529897 Supplier Invitrogen™ Supplier No. PA529897
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431. Predicted reactivity: Rhesus Monkey (97%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The proliferation and differentiation of B cells is dependent upon a B-cell antigen receptor (BCR) complex. Binding of antigens to specific B-cell receptors results in a tyrosine phosphorylation reaction through the BCR complex and leads to multiple signal transduction pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGBP1
Applications Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene IGBP1
Gene Accession No. P78318
Gene Alias AI115586; AI662457; alpha 4; ALPHA-4; Alpha4 phosphoprotein; AW208785; B cell signal transduction molecule alpha 4; bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ); B-cell signal transduction molecule alpha 4; C81413; CD79a-binding protein 1; IBP1; Igbp1; immunoglobulin (CD79A) binding protein 1; immunoglobulin binding protein 1; immunoglobulin-binding protein 1; Lymphocyte signal transduction molecule alpha 4; p52; Pc52; phosphoprotein, component, 52 kDa; protein alpha-4; Protein phosphatase 2/4/6 regulatory subunit; protein phosphatase 2A, regulatory subunit alpha-4; renal carcinoma antigen NY-REN-16; Tap42
Gene Symbols IGBP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IGBP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3476
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.