Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IGBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551429
Description
Immunogen sequence: AYPSLVAMAS QRQAKIQRYK QKKELEHRLS AMKSAVESGQ ADDERVREYY LLHLQRWIDI SLEEIESIDQ EIKILRERDS SREASTSNSS RQERPPVKPF ILTRNMAQAK VFGAGYPSLP Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 83%.
The proliferation and differentiation of B cells is dependent upon a B-cell antigen receptor (BCR) complex. Binding of antigens to specific B-cell receptors results in a tyrosine phosphorylation reaction through the BCR complex and leads to multiple signal transduction pathways.
Specifications
IGBP1 | |
Polyclonal | |
Unconjugated | |
IGBP1 | |
AI115586; AI662457; alpha 4; ALPHA-4; Alpha4 phosphoprotein; AW208785; B cell signal transduction molecule alpha 4; bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ); B-cell signal transduction molecule alpha 4; C81413; CD79a-binding protein 1; IBP1; Igbp1; immunoglobulin (CD79A) binding protein 1; immunoglobulin binding protein 1; immunoglobulin-binding protein 1; Lymphocyte signal transduction molecule alpha 4; p52; Pc52; phosphoprotein, component, 52 kDa; protein alpha-4; Protein phosphatase 2/4/6 regulatory subunit; protein phosphatase 2A, regulatory subunit alpha-4; renal carcinoma antigen NY-REN-16; Tap42 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3476 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P78318 | |
IGBP1 | |
Recombinant protein corresponding to Human IGBP1. Recombinant protein control fragment (Product #RP-91915). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction