Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Igbp1b Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570542
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 100%; Yeast: 77%.
Igbp1b gene ontology annotations related to this gene include regulation of signal transduction.
Specifications
Igbp1b | |
Polyclonal | |
Unconjugated | |
Igbp1b | |
alpha4-b; CD79a-binding protein 1b; Igbp1b; immunoglobulin (CD79A) binding protein 1b; immunoglobulin-binding protein 1b; RGD1560971 | |
Rabbit | |
Affinity chromatography | |
RUO | |
502919 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
Igbp1b | |
synthetic peptide directed towards the middle region of rat Igbp1b. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction