Learn More
IgD[a] Mouse anti-Mouse, Brilliant Violet 510, Clone: AMS 9.1, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743632
Description
The AMS 9.1 antibody reacts specifically with the Fc portion of mouse IgD of Igh C[a] /haplotype (eg, AKR, BALB/c, CBA, C3H/He, C58, DBA/1, DBA/2, NZB). It does not react with IgD of Igh-C[b] /or Igh-C[e] /haplotypes (eg, C57BL/6, C57BL/10, SJL, A). AMS 9.1 mAb is effective for detection of cell-surface Ig by immunofluorescent staining with flow cytometric analysis. It has been reported to stimulate B-cell proliferation.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
IgD[a] | |
AMS 9.1 | |
Brilliant Violet 510 | |
Igh-5a, Igh-5.4, Ighd, Immunoglobulin D | |
BALB/c mouse splenocytes | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.