Learn More
IgD[a] Mouse anti-Mouse, Brilliant Violet 711, Clone: AMS 9.1, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743635
Description
The AMS 9.1 antibody reacts specifically with the Fc portion of mouse IgD of Igh C[a] /haplotype (eg, AKR, BALB/c, CBA, C3H/He, C58, DBA/1, DBA/2, NZB). It does not react with IgD of Igh-C[b] /or Igh-C[e] /haplotypes (eg, C57BL/6, C57BL/10, SJL, A). AMS 9.1 mAb is effective for detection of cell-surface Ig by immunofluorescent staining with flow cytometric analysis. It has been reported to stimulate B-cell proliferation.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications
IgD[a] | |
AMS 9.1 | |
Brilliant Violet 711 | |
Igh-5a, Igh-5.4, Ighd, Immunoglobulin D | |
BALB/c mouse splenocytes | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.