Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Antibody (11-26c.2a), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP149000AF350
Description
IgD Monoclonal antibody specifically detects IgD in Mouse samples. It is validated for Flow CytometrySpecifications
IgD | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
11-26c.2a | |
Flow Cytometry | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
This antibody was generated by immunizing host with mouse IgD | |
0.5 mL | |
B Cell Development and Differentiation Markers, Immunology | |
3495 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction