Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Antibody (11-26c.2a), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP149000C
Description
IgD Monoclonal specifically detects IgD in Mouse samples. It is validated for Flow Cytometry.Specifications
IgD | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHD | |
This antibody was generated by immunizing host with mouse IgD | |
0.5 mL | |
3495 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry | |
11-26c.2a | |
Flow Cytometry | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction