Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Antibody (11-26C), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB9858H
Description
IgD Monoclonal antibody specifically detects IgD in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
IgD | |
Monoclonal | |
HRP | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
11-26C | |
Flow Cytometry, CyTOF-ready | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
Mouse IgD | |
0.1 mL | |
B Cell Development and Differentiation Markers, Immunology | |
3495 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction