Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD Antibody (IgD26) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB395425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395425 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395425 Supplier Novus Biologicals Supplier No. NBP2504370.025ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen IgD
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone IgD26
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, CyTOF-ready
Formulation PBS
Gene Alias IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Host Species Mouse
Immunogen IgD purified from human myeloma serum
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3495
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.