Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Antibody (IgD26), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250437MFV450
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow CytometrySpecifications
IgD | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
IgD26 | |
Flow Cytometry | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
IgD purified from human myeloma serum | |
0.1 mL | |
B Cell Development and Differentiation Markers, Immunology | |
3495 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction