Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Antibody (IgD26), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250437PEATT594
Description
IgD Monoclonal specifically detects IgD in Human samples. It is validated for Flow Cytometry.Specifications
IgD | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
IGHD | |
IgD purified from human myeloma serum | |
0.1 mL | |
3495 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
IgD26 | |
Flow Cytometry | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction