Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Antibody (IGHD/2730R), Novus Biologicals™

Rabbit Monoclonal Antibody
$495.50
Specifications
Antigen | IgD |
---|---|
Clone | IGHD/2730R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
IgD | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
B Cell Development and Differentiation Markers, Immunology | |
1mM PBS with 0.05% BSA | |
3495 | |
IgG κ | |
Protein A or G purified |
IGHD/2730R | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
Recombinant full-length human IgD protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title