Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Antibody (SR2250), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32189250UL
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
IgD | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR2250 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
A synthesized peptide derived from human IgD (Uniprot #: P01880 ) | |
50 μg | |
B Cell Development and Differentiation Markers, Immunology | |
3495 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction